Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000735.3 | NP_000726.1 | 3 | 336 | ||
NM_001252383.1 | NP_001239312.1 | 4 | 429 | |||
GenBank mRNA | AK311972.1 | - | 3 | 293 | ||
BC010957.1 | - | 3 | 299 | 4043527 | ||
BC020782.1 | - | 3 | 295 | 4732686 | ||
BC055080.1 | - | 3 | 297 | 4044086 | ||
BG571584.1 | - | 4 | 386 | |||
BG619154.1 | - | 3 | 337 | |||
CR457427.1 | - | 2 | 194 | |||
FJ624078.1 | - | 2 | 122 | |||
FJ624079.1 | - | 2 | 194 | |||
V00518.1 | - | 3 | 244 |