Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032385.4 | NP_115761.2 | 9 | 1377 | ||
XM_005268358.2 | XP_005268415.1 | 6 | 1022 | |||
XM_006714753.2 | XP_006714816.1 | 10 | 1752 | |||
XM_017008965.1 | XP_016864454.1 | 10 | 1777 | |||
XM_017008966.1 | XP_016864455.1 | 7 | 1052 | |||
GenBank mRNA | AF159165.1 | - | 1 | 1383 | ||
AK023820.1 | - | 8 | 2168 | |||
AK074245.1 | - | 3 | 522 | |||
AK127944.1 | - | 3 | 2711 | |||
BC004506.2 | - | 7 | 1074 | |||
BC007216.1 | - | 9 | 1345 | 2906244 |