Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R13L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142502.1 | NP_001135974.1 | 11 | 2275 | ||
NM_006663.3 | NP_006654.2 | 11 | 2273 | |||
XM_017026177.1 | XP_016881666.1 | 12 | 2331 | |||
XM_017026178.1 | XP_016881667.1 | 12 | 2391 | |||
GenBank mRNA | AF078036.1 | - | 5 | 1254 | ||
AF078037.1 | - | 4 | 1708 | |||
AJ888472.1 | - | 10 | 2196 | |||
AY869712.1 | - | 11 | 2254 | |||
BC001475.2 | - | 5 | 860 | |||
BC020589.1 | - | 4 | 514 | |||
BC032298.1 | - | 11 | 2262 | 4994121 | ||
BC064913.1 | - | 11 | 2238 | 6068949 | ||
DQ986418.1 | - | 4 | 933 |