Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001815.4 | NP_001806.2 | 7 | 833 | ||
NR_102333.2 | 8 | 1337 | ||||
GenBank mRNA | BC106728.1 | - | 7 | 771 | 40015072 | |
D90277.1 | - | 7 | 819 | |||
D90278.1 | - | 8 | 1310 | |||
E03348.1 | - | 7 | 819 | |||
E03349.1 | - | 9 | 1192 | |||
L00692.1 | - | 7 | 799 |