Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006667.4 | NP_006658.1 | 2 | 588 | ||
GenBank mRNA | AK313956.1 | - | 2 | 546 | ||
BC034238.1 | - | 2 | 522 | 3863377 | ||
CR456993.1 | - | 2 | 469 | |||
CX762728.1 | - | 2 | 585 | |||
Y12711.2 | - | 2 | 547 |