Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006785.3 | NP_006776.1 | 13 | 1772 | ||
NM_173844.2 | NP_776216.1 | 12 | 1739 | |||
GenBank mRNA | AB026118.1 | - | 12 | 1546 | ||
AF130356.2 | - | 13 | 1678 | |||
AF316597.1 | - | 13 | 1772 | |||
AK092004.1 | - | 2 | 1096 | |||
AK291386.1 | - | 13 | 1738 | |||
BC030143.2 | - | 12 | 1593 | 4765195 |