Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006694.3 | NP_006685.1 | 4 | 987 | ||
GenBank mRNA | AB016488.1 | - | 4 | 696 | ||
AF115850.2 | - | 4 | 685 | |||
AF131797.1 | - | 4 | 568 | |||
AF151056.1 | - | 3 | 743 | |||
AK312106.1 | - | 4 | 669 | |||
BC000499.1 | - | 4 | 709 | 2820861 | ||
BC000996.2 | - | 4 | 636 | 2901221 | ||
BC001363.1 | - | 4 | 674 | 3050800 | ||
BC001667.1 | - | 4 | 639 | 2823348 | ||
BC004239.1 | - | 4 | 696 | 3626047 | ||
BC019277.1 | - | 4 | 630 | 2823348 | ||
BT007285.1 | - | 4 | 264 | |||
CR456985.1 | - | 4 | 264 |