Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006699.3 | NP_006690.1 | 7 | 1714 | ||
XM_006710302.2 | XP_006710365.1 | 7 | 1722 | |||
XM_011540536.2 | XP_011538838.1 | 7 | 1784 | |||
GenBank mRNA | AF027156.1 | - | 7 | 1513 | ||
AK001970.1 | - | 3 | 191 | |||
BC063300.1 | - | 7 | 1712 | 5263292 |