Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289413.1 | NP_001276342.1 | 18 | 2575 | ||
NM_001318833.1 | NP_001305762.1 | 20 | 2782 | |||
NM_006709.4 | NP_006700.3 | 20 | 2532 | |||
NM_025256.6 | NP_079532.5 | 19 | 2430 | |||
XM_005248824.3 | XP_005248881.1 | 19 | 2673 | |||
XM_006714974.1 | XP_006715037.1 | 19 | 2698 | |||
XM_006714975.1 | XP_006715038.1 | 18 | 2596 | |||
XM_006714976.1 | XP_006715039.1 | 20 | 2536 | |||
XM_017010202.1 | XP_016865691.1 | 19 | 2434 | |||
GenBank mRNA | AB209433.2 | - | 19 | 2615 | ||
AJ315532.1 | - | 20 | 2502 | |||
AK092866.1 | - | 3 | 1447 | |||
AK302904.1 | - | 18 | 2575 | |||
BC002686.1 | - | 17 | 2002 | |||
BC009351.1 | - | 20 | 2499 | |||
BC018718.1 | - | 20 | 2490 | 4123580 | ||
BC020970.1 | - | 19 | 2467 | |||
X69838.1 | - | 16 | 1922 |