Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289413.1 | NP_001276342.1 | 6, 7 | 1043 | ||
NM_001318833.1 | NP_001305762.1 | 7, 8 | 1127 | |||
NM_006709.4 | NP_006700.3 | 7, 8 | 898 | |||
NM_025256.6 | NP_079532.5 | 7, 8 | 898 | |||
XM_005248824.3 | XP_005248881.1 | 6, 7 | 1039 | |||
XM_006714974.1 | XP_006715037.1 | 6, 7 | 1043 | |||
XM_006714975.1 | XP_006715038.1 | 6, 7 | 1043 | |||
XM_006714976.1 | XP_006715039.1 | 7, 8 | 881 | |||
XM_017010202.1 | XP_016865691.1 | 7, 8 | 881 | |||
GenBank mRNA | AB209433.2 | - | 6, 7 | 960 | ||
AJ315532.1 | - | 7, 8 | 868 | |||
AK302904.1 | - | 6, 7 | 1043 | |||
BC002686.1 | - | 4, 5 | 368 | |||
BC009351.1 | - | 7, 8 | 865 | |||
BC018718.1 | - | 7, 8 | 856 | 4123580 | ||
BC020970.1 | - | 6, 7 | 833 | |||
DA099526.1 | - | 4, 5 | 508 | |||
X69838.1 | - | 3, 4 | 288 |