Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318060.1 | NP_001304989.1 | 11 | 1892 | ||
NM_001318061.1 | NP_001304990.1 | 12 | 1907 | |||
NM_001318062.1 | NP_001304991.1 | 12 | 1938 | |||
NM_006716.3 | NP_006707.1 | 12 | 1999 | |||
GenBank mRNA | AB028069.1 | - | 12 | 2012 | ||
AB028070.1 | - | 12 | 1951 | |||
AF160249.1 | - | 12 | 1711 | |||
AF160876.1 | - | 12 | 1694 | |||
AK022969.1 | - | 1 | 359 | |||
AK292569.1 | - | 12 | 1741 | |||
AK303175.1 | - | 12 | 1603 | |||
BC036045.1 | - | 12 | 1805 | 5273059 | ||
BC047693.1 | - | 12 | 1680 | 5753567 |