Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282444.1 | NP_001269373.1 | 6, 7 | 1518 | ||
NM_001282445.1 | NP_001269374.1 | 5, 6 | 1196 | |||
NM_006795.3 | NP_006786.2 | 4, 5 | 1375 | |||
XM_011544739.1 | XP_011543041.1 | 5, 6 | 1193 | |||
GenBank mRNA | AF001434.1 | - | 4, 5 | 1326 | ||
AF099011.1 | - | 1317 | ||||
AK097842.1 | - | 6, 7 | 1483 | |||
AK098723.1 | - | 3, 4 | 646 | |||
AK124613.1 | - | 3, 4 | 2293 | |||
AK126581.1 | - | 2, 3 | 953 | |||
AK297654.1 | - | 5, 6 | 1054 | |||
AK299096.1 | - | 5, 6 | 1107 | |||
AK300326.1 | - | 5, 6 | 1156 | |||
AK302930.1 | - | 5, 6 | 1148 | |||
AK312316.1 | - | 4, 5 | 1228 | |||
AY007161.1 | - | 4, 5 | 1122 | |||
BC104799.1 | - | 4, 5 | 1267 | 8143802 | ||
BC104825.1 | - | 4, 5 | 1267 | 8143828 |