Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282652.1 | NP_001269581.1 | 5, 6 | 1360 | ||
NM_001282653.1 | NP_001269582.1 | 5, 6 | 741 | |||
NM_006819.2 | NP_006810.1 | 5, 6 | 813 | |||
GenBank mRNA | AK291555.1 | - | 5, 6 | 740 | ||
AK297319.1 | - | 5, 6 | 652 | |||
BC002987.1 | - | 5, 6 | 728 | 2823157 | ||
BC039299.1 | - | 5, 6 | 1360 | 4830381 | ||
BT020010.1 | - | 4, 5 | 666 | |||
BT020011.1 | - | 4, 5 | 666 | |||
CR536512.1 | - | 4, 5 | 666 | |||
FJ224350.1 | - | 5, 6 | 728 | |||
M86752.1 | - | 5, 6 | 728 |