Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006820.3 | NP_006811.2 | 5, 6 | 1077 | ||
XM_005270391.3 | XP_005270448.1 | 4, 5 | 584 | |||
XM_005270392.4 | XP_005270449.1 | 3, 4 | 364 | |||
XM_005270393.4 | XP_005270450.1 | 2, 3 | 286 | |||
XM_006710304.3 | XP_006710367.1 | 4, 5 | 391 | |||
XM_011540539.2 | XP_011538841.1 | 5, 6 | 915 | |||
XM_017000120.1 | XP_016855609.1 | 5, 6 | 585 | |||
GenBank mRNA | AB000115.1 | - | 5, 6 | 1001 | ||
AK225442.1 | - | 5, 6 | 978 | |||
AK293385.1 | - | 5, 6 | 676 | |||
AK303187.1 | - | 2, 3 | 278 | |||
AK316232.1 | - | 4, 5 | 526 | |||
AL832618.2 | - | 3, 4 | 3680 | |||
BC015932.2 | - | 5, 6 | 983 | 3909331 |