Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006826.3 | NP_006817.1 | 2 | 419 | ||
GenBank mRNA | AK297624.1 | - | 3 | 252 | ||
AK302208.1 | - | 2 | 125 | |||
AK309462.1 | - | 3 | 248 | |||
AK316158.1 | - | 3 | 247 | |||
BC001197.1 | - | 356 | ||||
BC050601.1 | - | 1 | 232 | 6501365 | ||
BC056867.1 | - | 2 | 354 | 6164592 | ||
BC093019.1 | - | 2 | 361 | 30520472 | ||
BT020014.1 | - | 1 | 222 | |||
HY044745.1 | - | 2 | 427 | |||
X56468.1 | - | 2 | 347 | |||
X57347.1 | - | 2 | 322 |