Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 41 | 5313 | ||
GenBank mRNA | BC153881.1 | - | 41 | 5313 | 8860118 | |
D86973.2 | - | 41 | 5313 | |||
U77700.1 | - | 19 | 2778 | |||
U88836.1 | - | 2 | 271 |