Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317182.1 | NP_001304111.1 | 7, 8 | 930 | ||
NM_001317183.1 | NP_001304112.1 | 6, 7 | 930 | |||
NM_006838.3 | NP_006829.1 | 7, 8 | 999 | |||
NR_133673.1 | 7, 8 | 999 | ||||
XM_005268583.3 | XP_005268640.1 | 7, 8 | 918 | |||
GenBank mRNA | AK055692.1 | - | 721 | |||
AK091730.1 | - | 6, 7 | 806 | |||
AK125296.1 | - | 7, 8 | 1525 | |||
AK300836.1 | - | 7, 8 | 895 | |||
AK315559.1 | - | 7, 8 | 936 | |||
BC013782.1 | - | 7, 8 | 877 | 4340717 | ||
BC016008.1 | - | 2, 3 | 192 | |||
U13261.1 | - | 7, 8 | 899 |