Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005094.3 | NP_005085.2 | 13 | 2118 | ||
XM_017014222.1 | XP_016869711.1 | 14 | 1991 | |||
GenBank mRNA | AK000722.1 | - | 4 | 481 | ||
AK027504.1 | - | 12 | 1698 | |||
AK074831.1 | - | 10 | 1166 | |||
AK290222.1 | - | 13 | 2118 | |||
AK314681.1 | - | 12 | 2138 | |||
BC004268.1 | - | 6 | 678 | |||
BC009959.2 | - | 6 | 731 | |||
BC065003.1 | - | 13 | 2117 | 6023438 |