Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100603.1 | NP_001094073.1 | 4 | 532 | ||
NM_006854.3 | NP_006845.1 | 5 | 785 | |||
GenBank mRNA | AK093917.1 | - | 3 | 1084 | ||
AK315702.1 | - | 5 | 757 | |||
AL547016.3 | - | 5 | 762 | |||
BC008081.2 | - | 5 | 689 | 2966703 | ||
BC012994.2 | - | 5 | 720 | 3604062 | ||
BC071982.1 | - | 4 | 486 | 5477638 | ||
M88458.1 | - | 5 | 662 | |||
X63745.1 | - | 5 | 636 |