Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083963.1 | NP_001077432.1 | 9 | 1414 | ||
NM_001083964.1 | NP_001077433.1 | 9 | 1304 | |||
NM_001083965.1 | NP_001077434.1 | 9 | 1439 | |||
NM_006862.3 | NP_006853.2 | 9 | 1439 | |||
XM_005244856.4 | XP_005244913.1 | 9 | 1205 | |||
XM_017000122.1 | XP_016855611.1 | 9 | 1481 | |||
XM_017000123.1 | XP_016855612.1 | 9 | 1481 | |||
XM_017000124.1 | XP_016855613.1 | 9 | 1380 | |||
XM_017000125.1 | XP_016855614.1 | 9 | 1409 | |||
XM_017000126.1 | XP_016855615.1 | 9 | 1380 | |||
XM_017000127.1 | XP_016855616.1 | 9 | 1409 | |||
XM_017000128.1 | XP_016855617.1 | 9 | 1334 | |||
XM_017000129.1 | XP_016855618.1 | 9 | 1299 | |||
XM_017000130.1 | XP_016855619.1 | 9 | 1273 | |||
GenBank mRNA | AF119121.2 | - | 9 | 1427 | ||
AF227192.1 | - | 9 | 1427 | |||
AK056402.1 | - | 9 | 1362 | |||
AK225160.1 | - | 9 | 1293 | |||
AK226092.1 | - | 9 | 1352 | |||
AK295948.1 | - | 9 | 1210 | |||
AK303063.1 | - | 9 | 1335 | |||
BC032690.1 | - | 9 | 1346 |