Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 22 | 2719 | ||
XM_005250760.3 | XP_005250817.1 | 21 | 2680 | |||
XM_005250761.3 | XP_005250818.1 | 22 | 2760 | |||
XM_011516802.2 | XP_011515104.1 | 20 | 2408 | |||
XM_017012993.1 | XP_016868482.1 | 21 | 2672 | |||
XM_017012994.1 | XP_016868483.1 | 20 | 2598 | |||
XM_017012995.1 | XP_016868484.1 | 20 | 2541 | |||
XM_017012996.1 | XP_016868485.1 | 19 | 2459 | |||
XM_017012997.1 | XP_016868486.1 | 18 | 2366 | |||
GenBank mRNA | AK292800.1 | - | 22 | 2734 | ||
AL136739.1 | - | 20 | 2449 | |||
AY043361.1 | - | 21 | 2524 | |||
AY345857.1 | - | 22 | 2753 | |||
BC015380.2 | - | 5 | 573 | |||
BC036065.1 | - | 22 | 2723 | 5296005 | ||
U96113.1 | - | 15 | 1873 |