Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270419.1 | NP_001257348.1 | 7 | 749 | ||
NM_181581.2 | NP_853559.1 | 7 | 899 | |||
NR_073002.1 | 7 | 999 | ||||
NR_073003.1 | 6 | 781 | ||||
NR_073004.1 | 6 | 738 | ||||
NR_073005.1 | 6 | 631 | ||||
XM_005250125.4 | XP_005250182.1 | 6 | 761 | |||
XM_006715844.2 | XP_006715907.1 | 5 | 637 | |||
XM_006715845.3 | XP_006715908.1 | 4 | 548 | |||
XM_011515766.2 | XP_011514068.1 | 7 | 893 | |||
XM_011515767.2 | XP_011514069.1 | 6 | 643 | |||
XM_017011708.1 | XP_016867197.1 | 7 | 914 | |||
XM_017011709.1 | XP_016867198.1 | 7 | 894 | |||
XM_017011710.1 | XP_016867199.1 | 7 | 748 | |||
XM_017011711.1 | XP_016867200.1 | 6 | 676 | |||
XM_017011712.1 | XP_016867201.1 | 6 | 660 | |||
XM_017011713.1 | XP_016867202.1 | 6 | 730 | |||
GenBank mRNA | AK127802.1 | - | 3 | 3823 | ||
AK294095.1 | - | 6 | 737 | |||
AK297195.1 | - | 7 | 968 | |||
AK308472.1 | - | 2 | 475 | |||
AW954755.1 | - | 2 | 184 | |||
BC111774.1 | - | 5 | 1117 | 40033792 | ||
BX357552.2 | - | 3 | 214 | |||
BX395100.2 | - | 7 | 708 | |||
CX787147.1 | - | 5 | 468 | |||
U62767.1 | - | 7 | 697 |