Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100397.1 | NP_001093867.1 | 2 | 418 | ||
NM_001282899.1 | NP_001269828.1 | 2 | 289 | |||
NM_001282900.1 | NP_001269829.1 | 2 | 289 | |||
NM_007023.3 | NP_008954.2 | 5 | 659 | |||
XM_005246246.2 | XP_005246303.1 | 5 | 526 | |||
XM_006712205.3 | XP_006712268.1 | 5 | 531 | |||
XM_011510505.2 | XP_011508807.1 | 5 | 526 | |||
XM_017003196.1 | XP_016858685.1 | 5 | 526 | |||
XM_017003197.1 | XP_016858686.1 | 2 | 229 | |||
GenBank mRNA | AB027471.1 | - | 5 | 529 | ||
AB209681.1 | - | 5 | 588 | |||
AK294445.1 | - | 2 | 138 | |||
AK294997.1 | - | 5 | 624 | |||
AK295002.1 | - | 2 | 224 | |||
AK296340.1 | - | 2 | 284 | |||
AK304278.1 | - | 2 | 178 | |||
AK311165.1 | - | 2 | 111 | |||
AK311500.1 | - | 2 | 418 | |||
AK311697.1 | - | 2 | 395 | |||
AK313084.1 | - | 5 | 528 | |||
AK316014.1 | - | 2 | 253 | |||
AK316072.1 | - | 2 | 185 | |||
BC024004.2 | - | 5 | 498 | 4823935 | ||
BC040183.2 | - | 2 | 419 | 4801092 | ||
DC335305.1 | - | 2 | 289 | |||
KJ534924.1 | - | 5 | 436 | |||
KJ534925.1 | - | 5 | 472 | |||
KJ534978.1 | - | 5 | 472 | |||
U78516.1 | - | 5 | 580 |