Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316897.1 | NP_001303826.1 | 4 | 523 | ||
NM_001316898.1 | NP_001303827.1 | 4 | 516 | |||
NM_001316900.1 | NP_001303829.1 | 4 | 523 | |||
NM_018956.4 | NP_061829.3 | 4 | 523 | |||
NR_133631.1 | 3 | 313 | ||||
XM_017014231.1 | XP_016869720.1 | 4 | 519 | |||
GenBank mRNA | AJ011375.1 | - | 4 | 511 | ||
AK075259.1 | - | 4 | 516 | |||
AK130389.1 | - | 1 | 596 | |||
BC012940.1 | - | 4 | 473 | 4478288 | ||
CK906174.1 | - | 2 | 204 | |||
CK906175.1 | - | 4 | 469 | |||
CR457210.1 | - | 3 | 362 | |||
DA134573.1 | - | 4 | 516 | |||
DA281620.1 | - | 3 | 267 |