Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291296.1 | NP_001278225.1 | 5 | 596 | ||
NM_007059.3 | NP_008990.2 | 7 | 764 | |||
NR_111923.1 | 8 | 851 | ||||
XM_011526399.2 | XP_011524701.1 | 10 | 2269 | |||
XM_017026226.1 | XP_016881715.1 | 10 | 1114 | |||
XM_017026227.1 | XP_016881716.1 | 9 | 1190 | |||
XM_017026228.1 | XP_016881717.1 | 10 | 1114 | |||
GenBank mRNA | AF105369.1 | - | 7 | 696 | ||
AK023996.1 | - | 7 | 719 | |||
AK225419.1 | - | 7 | 708 | |||
AK298672.1 | - | 8 | 749 | |||
BC009249.2 | - | 7 | 679 | 3029623 | ||
DR759729.1 | - | 5 | 508 |