Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 19, 20 | 3109 | ||
XM_005263862.3 | XP_005263919.1 | 19, 20 | 3398 | |||
XM_005263863.4 | XP_005263920.1 | 19, 20 | 3185 | |||
XM_005263864.3 | XP_005263921.1 | 19, 20 | 3350 | |||
XM_011510507.1 | XP_011508809.1 | 21, 22 | 3240 | |||
XM_017003198.1 | XP_016858687.1 | 18, 19 | 3305 | |||
XM_017003199.1 | XP_016858688.1 | 18, 19 | 3258 | |||
GenBank mRNA | AB449889.1 | - | 19, 20 | 2978 | ||
AK292826.1 | - | 17, 18 | 2742 | |||
CD722231.1 | - | 2, 3 | 177 |