Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDB3 SIRNA Assays ›
Gene Symbol:
LDB3
Gene Name:
LIM domain binding 3
Gene Aliases:
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP
Chromosome Location:
Chr.10: 86664785 - 86736072 on Build GRCh38
UniGene ID:
Hs.657271
Species:
Human
siRNA ID
HSS174126
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080114.1
NP_001073583.1
2
176
NM_001080115.1
NP_001073584.1
2
176
NM_001080116.1
NP_001073585.1
2
281
NM_001171610.1
NP_001165081.1
3
298
NM_001171611.1
NP_001165082.1
2
176
NM_007078.2
NP_009009.1
2
176
XM_005269464.4
XP_005269521.1
3
345
XM_005269466.4
XP_005269523.1
3
344
XM_005269468.4
XP_005269525.1
3
341
XM_011539184.2
XP_011537486.1
3
346
XM_011539185.2
XP_011537487.1
3
2240
XM_011539186.2
XP_011537488.1
3
346
XM_011539187.2
XP_011537489.1
3
345
XM_011539188.2
XP_011537490.1
3
346
XM_011539189.1
XP_011537491.1
3
240
XM_011539190.1
XP_011537492.1
2
176
XM_011539191.2
XP_011537493.1
3
345
XM_011539193.1
XP_011537495.1
3
240
XM_011539194.1
XP_011537496.1
3
240
XM_011539195.2
XP_011537497.1
3
342
XM_017015606.1
XP_016871095.1
3
2240
XM_017015608.1
XP_016871097.1
3
2240
XM_017015609.1
XP_016871098.1
3
240
GenBank mRNA
AB014513.1
-
2
174
AF276807.1
-
2
153
AF276808.1
-
2
153
AF276809.1
-
2
153
AJ133766.1
-
2
232
AJ133767.1
-
2
153
AJ133768.1
-
2
153
AK294696.1
-
3
233
AK297871.1
-
3
132
AK300692.1
-
3
189
AK304760.1
-
3
240
AL601484.1
-
2
188
BC010929.2
-
2
178
DA898241.1
-
2
281
DC357269.1
-
3
298
KF772970.1
-
2
153
KU178543.1
-
2
153
KU178544.1
-
2
153
KU178545.1
-
2
153
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
sarcomere organization
protein kinase C binding
protein binding
cytoskeletal protein binding
zinc ion binding
muscle alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0