Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007176.3 | NP_009107.1 | 2 | 493 | ||
GenBank mRNA | AF134159.2 | - | 2 | 113 | ||
AF136971.1 | - | 2 | 153 | |||
AF161406.1 | - | 2 | 153 | |||
AK311764.1 | - | 2 | 133 | |||
AL136658.1 | - | 2 | 151 | |||
BC002444.2 | - | 2 | 160 | 3347242 | ||
BX248022.1 | - | 2 | 487 | |||
CR533443.1 | - | 1 | 41 |