Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007085.4 | NP_009016.1 | 5, 6 | 507 | ||
GenBank mRNA | AK289388.1 | - | 5, 6 | 428 | ||
AK291138.1 | - | 5, 6 | 429 | |||
AK300356.1 | - | 4, 5 | 322 | |||
AK300436.1 | - | 5, 6 | 358 | |||
AK312261.1 | - | 5, 6 | 434 | |||
BC000055.2 | - | 5, 6 | 414 | 3505833 | ||
BX647421.1 | - | 5, 6 | 432 | |||
DB289587.1 | - | 5, 6 | 507 | |||
U06863.1 | - | 5, 6 | 423 |