Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145099.1 | NP_001138571.1 | 2 | 333 | ||
NM_017585.3 | NP_060055.2 | 2 | 333 | |||
XM_011518189.2 | XP_011516491.2 | 2 | 306 | |||
XM_017014236.1 | XP_016869725.1 | 2 | 315 | |||
XM_017014237.1 | XP_016869726.1 | 2 | 306 | |||
XM_017014238.1 | XP_016869727.1 | 2 | 306 | |||
GenBank mRNA | AB209108.1 | - | 1 | 598 | ||
AJ011372.2 | - | 2 | 275 | |||
AK074836.1 | - | 2 | 283 | |||
AK074927.1 | - | 2 | 316 | |||
AK096630.1 | - | 1 | 836 | |||
AK222919.1 | - | 2 | 278 | |||
AK225106.1 | - | 2 | 275 | |||
AK294974.1 | - | 2 | 253 | |||
BC013740.1 | - | 2 | 267 | 4449594 | ||
DC305025.1 | - | 2 | 333 | |||
Y17803.1 | - | 2 | 284 |