Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007202.3 | NP_009133.2 | 4 | 842 | ||
XM_006721432.3 | XP_006721495.1 | 4 | 837 | |||
XM_017024071.1 | XP_016879560.1 | 4 | 837 | |||
GenBank mRNA | AF037439.1 | - | 4 | 802 | ||
AK295180.1 | - | 4 | 784 | |||
AK308297.1 | - | 4 | 689 | |||
AK312438.1 | - | 4 | 795 | |||
BC017055.1 | - | 4 | 798 | 3882941 | ||
DQ270159.1 | - | 4 | 665 | |||
KU178546.1 | - | 4 | 665 | |||
KU178547.1 | - | 4 | 665 | |||
KU178548.1 | - | 3 | 482 |