Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145239.2 | NP_660282.2 | 3, 4 | 1303 | ||
XM_017022888.1 | XP_016878377.1 | 3, 4 | 1232 | |||
GenBank mRNA | AK098379.1 | - | 4, 5 | 1015 | ||
AK225785.1 | - | 3, 4 | 1198 | |||
AL834185.1 | - | 3, 4 | 1144 | |||
BC011405.1 | - | 3, 4 | 1194 | 3868330 | ||
BC053594.1 | - | 3, 4 | 1122 | 5729288 |