Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007225.2 | NP_009156.2 | 2 | 878 | ||
GenBank mRNA | AB032985.1 | - | 1 | 502 | ||
AF043468.1 | - | 1 | 357 | |||
AK308928.1 | - | 2 | 692 | |||
AL834282.1 | - | 2 | 956 | |||
AY358526.1 | - | 2 | 648 | |||
BC022541.1 | - | 2 | 697 | 4794968 |