Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145211.2 | NP_001138683.1 | 9 | 1475 | ||
NM_001145212.2 | NP_001138684.1 | 6 | 1348 | |||
NM_007256.4 | NP_009187.1 | 9 | 1780 | |||
XM_017017157.1 | XP_016872646.1 | 9 | 1398 | |||
GenBank mRNA | AB020687.1 | - | 9 | 1651 | ||
AB026256.1 | - | 9 | 1563 | |||
AF205073.1 | - | 9 | 1497 | |||
AK290234.1 | - | 9 | 1662 | |||
AK294503.1 | - | 9 | 1460 | |||
AK300134.1 | - | 6 | 1220 | |||
AK302492.1 | - | 7 | 1425 | |||
AK304783.1 | - | 4 | 932 | |||
AL117465.1 | - | 5 | 952 | |||
BC041095.1 | - | 9 | 1649 | 5752976 |