Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145211.2 | NP_001138683.1 | 6 | 831 | ||
NM_001145212.2 | NP_001138684.1 | 3 | 704 | |||
NM_007256.4 | NP_009187.1 | 6 | 1136 | |||
XM_017017157.1 | XP_016872646.1 | 6 | 754 | |||
GenBank mRNA | AB020687.1 | - | 6 | 1007 | ||
AB026256.1 | - | 6 | 919 | |||
AF205073.1 | - | 6 | 853 | |||
AK290234.1 | - | 6 | 1018 | |||
AK294503.1 | - | 6 | 816 | |||
AK300134.1 | - | 3 | 576 | |||
AK302492.1 | - | 4 | 781 | |||
BC041095.1 | - | 6 | 1005 | 5752976 |