Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080668.3 | NP_542399.1 | 2 | 282 | ||
XM_005273733.3 | XP_005273790.1 | 2 | 277 | |||
XM_005273734.3 | XP_005273791.1 | 2 | 277 | |||
XM_011544743.2 | XP_011543045.1 | 2 | 277 | |||
XM_011544747.2 | XP_011543049.1 | 2 | 268 | |||
XM_011544748.2 | XP_011543050.1 | 2 | 277 | |||
XM_011544749.2 | XP_011543051.1 | 2 | 277 | |||
XM_017017158.1 | XP_016872647.1 | 2 | 277 | |||
GenBank mRNA | AK291490.1 | - | 2 | 206 | ||
BC011000.2 | - | 2 | 190 | 3938081 | ||
CN332525.1 | - | 2 | 281 |