Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052880.4 | NP_443112.2 | 5 | 759 | ||
GenBank mRNA | AF528079.1 | - | 5 | 704 | ||
AF528080.1 | - | 5 | 704 | |||
AK074688.1 | - | 5 | 705 | |||
AK093768.1 | - | 2 | 1319 | |||
AK226089.1 | - | 5 | 639 | |||
AK300990.1 | - | 6 | 617 | |||
BC011049.2 | - | 5 | 704 | 4180134 | ||
BC041903.1 | - | 3 | 1056 | |||
BE299406.1 | - | 3 | 387 | |||
BQ718653.1 | - | 5 | 737 | |||
CR456340.1 | - | 5 | 599 | |||
GU135609.1 | - | 3 | 383 |