Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000751.2 | NP_000742.1 | 4 | 321 | ||
NM_001256657.1 | NP_001243586.1 | 3 | 276 | |||
NM_001311195.1 | NP_001298124.1 | 4 | 321 | |||
NM_001311196.1 | NP_001298125.1 | 4 | 321 | |||
XM_011510524.2 | XP_011508826.1 | 4 | 293 | |||
GenBank mRNA | AK291526.1 | - | 4 | 293 | ||
AK296704.1 | - | 4 | 293 | |||
AK300109.1 | - | 3 | 276 | |||
AK304890.1 | - | 4 | 293 | |||
AK315297.1 | - | 4 | 293 | |||
BC093923.1 | - | 4 | 297 | 7939768 | ||
BC093925.1 | - | 4 | 297 | 7939770 | ||
CD013895.1 | - | 4 | 266 | |||
X55019.1 | - | 4 | 269 |