Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301627.1 | NP_001288556.1 | 4 | 765 | ||
NM_052917.3 | NP_443149.2 | 6 | 1243 | |||
XM_011510537.2 | XP_011508839.1 | 6 | 1238 | |||
XM_011510538.2 | XP_011508840.1 | 4 | 650 | |||
XM_017003258.1 | XP_016858747.1 | 6 | 1092 | |||
XM_017003259.1 | XP_016858748.1 | 5 | 817 | |||
XM_017003260.1 | XP_016858749.1 | 6 | 1238 | |||
XM_017003261.1 | XP_016858750.1 | 6 | 1102 | |||
XM_017003262.1 | XP_016858751.1 | 4 | 508 | |||
XM_017003263.1 | XP_016858752.1 | 4 | 650 | |||
GenBank mRNA | AB067505.1 | - | 4 | 543 | ||
AB078142.1 | - | 4 | 661 | |||
AJ505991.2 | - | 4 | 661 | |||
AK131195.1 | - | 6 | 1188 | |||
AK308675.1 | - | 4 | 650 | |||
BC101031.1 | - | 4 | 661 | 40007710 | ||
BC101032.2 | - | 4 | 661 | 40007711 | ||
BC101033.1 | - | 4 | 661 | 40007712 | ||
BC101034.2 | - | 4 | 661 | 40007713 | ||
KJ534843.1 | - | 4 | 661 | |||
KJ535058.1 | - | 3 | 492 |