Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052928.2 | NP_443160.2 | 7 | 1934 | ||
XM_011523632.2 | XP_011521934.1 | 7 | 1915 | |||
XM_011523633.2 | XP_011521935.1 | 7 | 1915 | |||
XM_017024121.1 | XP_016879610.1 | 8 | 2179 | |||
GenBank mRNA | AB067523.1 | - | 3 | 1178 | ||
AK057769.1 | - | 3 | 366 | |||
AK092149.1 | - | 3 | 839 | |||
AK095369.1 | - | 7 | 1934 | |||
BC035077.2 | - | 7 | 1853 | 5260399 |