Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030968.3 | NP_112230.1 | 4 | 762 | ||
NM_153372.1 | NP_699203.1 | 4 | 498 | |||
NM_198593.3 | NP_940995.1 | 4 | 578 | |||
NM_198594.2 | NP_940996.1 | 3 | 593 | |||
NR_049769.1 | 4 | 616 | ||||
XM_006721663.1 | XP_006721726.1 | 3 | 623 | |||
XM_006721664.1 | XP_006721727.1 | 6 | 1210 | |||
XM_006721666.1 | XP_006721729.1 | 5 | 826 | |||
XM_017024126.1 | XP_016879615.1 | 3 | 867 | |||
GenBank mRNA | AF232905.1 | - | 3 | 705 | ||
AF329840.1 | - | 4 | 499 | |||
AJ272138.1 | - | 3 | 705 | |||
AK055541.1 | - | 3 | 408 | |||
AK075175.1 | - | 2 | 103 | |||
AK172767.1 | - | 4 | 599 | |||
AK292034.1 | - | 4 | 616 | |||
AY358424.1 | - | 4 | 728 | |||
BC021553.1 | - | 4 | 384 | 4622952 | ||
DA877694.1 | - | 4 | 498 |