Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242463.1 | NP_001229392.1 | 5 | 650 | ||
NM_058229.3 | NP_478136.1 | 7 | 929 | |||
NM_148177.2 | NP_680482.1 | 4 | 564 | |||
GenBank mRNA | AJ420108.1 | - | 7 | 779 | ||
AK056986.1 | - | 7 | 882 | |||
AY059629.1 | - | 7 | 690 | |||
BC024030.2 | - | 4 | 564 | 4825638 | ||
BC100020.1 | - | 6 | 520 | |||
BC120963.1 | - | 4 | 558 | 40115317 | ||
BC120964.1 | - | 4 | 558 | 40115318 | ||
EF143260.1 | - | 5 | 603 |