Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178450.4 | NP_848545.1 | 2 | 669 | ||
XM_011543128.2 | XP_011541430.1 | 2 | 611 | |||
XM_011543129.1 | XP_011541431.1 | 2 | 453 | |||
XM_011543130.1 | XP_011541432.1 | 2 | 358 | |||
XM_011543131.2 | XP_011541433.1 | 2 | 587 | |||
XM_011543132.1 | XP_011541434.1 | 2 | 1405 | |||
XM_017009014.1 | XP_016864503.1 | 2 | 1062 | |||
XM_017009015.1 | XP_016864504.1 | 1 | 210 | |||
GenBank mRNA | AF055007.1 | - | 2 | 325 | ||
AK290129.1 | - | 2 | 453 | |||
BC047569.1 | - | 2 | 419 | 4819831 | ||
BC146948.1 | - | 2 | 238 | 9056876 | ||
BC146964.1 | - | 2 | 238 | 9056892 | ||
BG719261.1 | - | 2 | 677 |