Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178450.4 | NP_848545.1 | 3, 4 | 904 | ||
GenBank mRNA | AF055007.1 | - | 3, 4 | 560 | ||
AK290129.1 | - | 3, 4 | 688 | |||
BC047569.1 | - | 3, 4 | 654 | 4819831 |