Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146032.1 | NP_001139504.1 | 17 | 1431 | ||
NM_138782.2 | NP_620137.2 | 18 | 1530 | |||
XM_017009016.1 | XP_016864505.1 | 17 | 1409 | |||
XM_017009017.1 | XP_016864506.1 | 18 | 1524 | |||
XM_017009018.1 | XP_016864507.1 | 18 | 1524 | |||
XM_017009019.1 | XP_016864508.1 | 18 | 1524 | |||
GenBank mRNA | AK291782.1 | - | 18 | 1460 | ||
AK293796.1 | - | 2 | 173 | |||
AK295141.1 | - | 17 | 1341 | |||
AL831971.1 | - | 18 | 1439 | |||
AL833313.1 | - | 15 | 1134 | |||
BC137070.1 | - | 18 | 1530 | 9021067 |