Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146032.1 | NP_001139504.1 | 22 | 2076 | ||
NM_138782.2 | NP_620137.2 | 23 | 2175 | |||
XM_017009016.1 | XP_016864505.1 | 22 | 2054 | |||
GenBank mRNA | AK291782.1 | - | 23 | 2105 | ||
AK293796.1 | - | 7 | 818 | |||
AK295141.1 | - | 22 | 1986 | |||
AL831971.1 | - | 23 | 2084 | |||
AL832928.1 | - | 5 | 516 | |||
AL833313.1 | - | 20 | 1774 | |||
BC137070.1 | - | 23 | 2175 | 9021067 |