Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172630.1 | NP_001166101.1 | 21 | 3711 | ||
NM_052948.3 | NP_443180.2 | 21 | 3410 | |||
XM_005258488.1 | XP_005258545.1 | 21 | 3889 | |||
XM_006722999.1 | XP_006723062.1 | 21 | 3922 | |||
XM_006723003.2 | XP_006723066.1 | 6 | 2422 | |||
XM_011526416.1 | XP_011524718.1 | 20 | 3777 | |||
XM_011526417.1 | XP_011524719.1 | 20 | 3750 | |||
XM_011526418.1 | XP_011524720.1 | 21 | 3696 | |||
XM_011526419.1 | XP_011524721.1 | 21 | 3663 | |||
XM_011526420.1 | XP_011524722.1 | 20 | 3828 | |||
GenBank mRNA | AK096338.1 | - | 21 | 3351 | ||
AK127255.1 | - | 21 | 3710 | |||
AL137579.1 | - | 8 | 2475 | |||
BC014084.2 | - | 1 | 773 |