Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286812.1 | NP_001273741.1 | 3 | 490 | ||
NM_198273.2 | NP_938014.1 | 3 | 549 | |||
XM_005248421.4 | XP_005248478.1 | 3 | 1014 | |||
GenBank mRNA | AK291478.1 | - | 3 | 546 | ||
AL137315.1 | - | 1 | 46 | |||
AL832721.1 | - | 3 | 530 | |||
BC058027.1 | - | 3 | 490 | |||
BC136740.1 | - | 3 | 441 | 9020730 | ||
BC136741.1 | - | 3 | 441 | 9020731 | ||
BP873090.1 | - | 3 | 545 | |||
BX537972.1 | - | 3 | 519 | |||
DN994993.1 | - | 3 | 558 |