Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012426.1 | NP_001012426.1 | 2 | 492 | ||
NM_001012427.1 | NP_001012427.1 | 2 | 492 | |||
NM_138457.2 | NP_612466.1 | 2 | 492 | |||
XM_006714991.3 | XP_006715054.1 | 2 | 297 | |||
XM_011514289.2 | XP_011512591.1 | 2 | 369 | |||
XM_011514290.2 | XP_011512592.1 | 2 | 370 | |||
XM_011514291.2 | XP_011512593.1 | 2 | 368 | |||
XM_011514292.2 | XP_011512594.1 | 2 | 370 | |||
XM_017010233.1 | XP_016865722.1 | 2 | 1501 | |||
XM_017010234.1 | XP_016865723.1 | 2 | 369 | |||
GenBank mRNA | AB080747.1 | - | 1 | 46 | ||
BC040962.1 | - | 2 | 492 | 5527139 | ||
BC052803.1 | - | 2 | 270 | 6420057 |