Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256476.1 | NP_001243405.1 | 1 | 161 | ||
NM_138458.3 | NP_612467.1 | 1 | 161 | |||
NR_046234.1 | 1 | 147 | ||||
GenBank mRNA | AK056303.1 | - | 1 | 125 | ||
AK058090.1 | - | 1 | 147 | |||
AL834142.1 | - | 1 | 110 | |||
AY541587.1 | - | 1 | 136 | |||
BC014022.2 | - | 1 | 83 | 4309224 | ||
BC020271.1 | - | 1 | 115 | |||
BC036904.1 | - | 1 | 84 | |||
BC066657.1 | - | 1 | 111 | 6164689 | ||
BP271204.1 | - | 1 | 161 |